The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017385.1 | NP_001017385.1 | 4 - 5 | 698 | 87 |
GenBank mRNA | BC092600.1 | - | 4 - 5 | 698 | 87 |