The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100634.2 | NP_001094104.1 | 11 - 12 | 1280 | 69 |
XM_006229218.2 | XP_006229280.1 | 10 - 11 | 1406 | 69 | |
XM_006229219.1 | XP_006229281.1 | 11 - 12 | 1319 | 69 | |
GenBank mRNA | CB609064.1 | - | 4 - 5 | 395 | 69 |
X53501.1 | - | 11 - 12 | 1280 | 69 |