The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100634.2 | NP_001094104.1 | 9 - 10 | 1050 | 63 |
XM_006229218.2 | XP_006229280.1 | 8 - 9 | 1176 | 63 | |
XM_006229219.1 | XP_006229281.1 | 9 - 10 | 1089 | 63 | |
GenBank mRNA | CB609064.1 | - | 2 - 3 | 165 | 63 |
X53501.1 | - | 9 - 10 | 1050 | 63 |