Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trappc6a
Entrez Gene ID
680465
Gene Name
trafficking protein particle complex 6A
Gene Aliases
RGD1309160
Chromosome Location
Chr.1: 80417514 - 80424429 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109410.1 NP_001102880.1 2 - 3 179 145

View Full Product Details