Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kptn
Entrez Gene ID
308107
Gene Name
kaptin (actin binding protein)
Gene Aliases
-
Chromosome Location
Chr.1: 78015714 - 78016508 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC166768.1 - - 493 79

View Full Product Details