Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem50b
Entrez Gene ID
360698
Gene Name
transmembrane protein 50B
Gene Aliases
RGD1307561
Chromosome Location
Chr.11: 31719441 - 31752277 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025014.1 NP_001020185.1 5 - 6 533 135
XM_006248066.2 XP_006248128.1 6 - 7 701 135
XM_006248067.2 XP_006248129.1 6 - 7 787 135
XM_006248068.2 XP_006248130.1 5 - 6 653 135
XM_006248069.2 XP_006248131.1 5 - 6 539 135
GenBank mRNA BC091349.1 - 5 - 6 533 135

View Full Product Details