Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm2d3
Entrez Gene ID
292995
Gene Name
TM2 domain containing 3
Gene Aliases
RGD1564929
Chromosome Location
Chr.1: 126573474 - 126595778 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106267.1 NP_001099737.1 - 1082 81

View Full Product Details