Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snrpa1
Entrez Gene ID
100361269
Gene Name
small nuclear ribonucleoprotein polypeptide A'
Gene Aliases
-
Chromosome Location
Chr.1: 126961389 - 126974453 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271323.1 NP_001258252.1 4 - 5 428 240
XM_006229323.2 XP_006229385.1 4 - 5 587 240
GenBank mRNA FQ219673.1 - 4 - 5 412 160
FQ231357.1 - 4 - 5 428 160
FQ232647.1 - 4 - 5 413 240

View Full Product Details