Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otud7a
Entrez Gene ID
309252
Gene Name
OTU deubiquitinase 7A
Gene Aliases
RGD1563721
Chromosome Location
Chr.1: 124507781 - 124655437 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759566.1 XP_008757788.1 10 - 11 1751 65
XM_008759567.1 XP_008757789.1 10 - 11 2032 65
XM_008759568.1 XP_008757790.1 10 - 11 1748 65

View Full Product Details