Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcee
Entrez Gene ID
293829
Gene Name
methylmalonyl CoA epimerase
Gene Aliases
-
Chromosome Location
Chr.1: 125229487 - 125252692 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106341.1 NP_001099811.1 2 - 3 515 67

View Full Product Details