Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lysmd4
Entrez Gene ID
681647
Gene Name
LysM, putative peptidoglycan-binding, domain containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 128199113 - 128204989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748880.3 XP_003748928.1 2 - 3 207 116
XM_008759576.1 XP_008757798.1 2 - 3 210 116

View Full Product Details