The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012158.1 | NP_001012158.1 | 5 - 6 | 877 | 66 |
GenBank mRNA | AB296367.1 | - | 4 - 5 | 786 | 66 |
AB296368.1 | - | 3 - 4 | 642 | 66 | |
BC087079.1 | - | 5 - 6 | 877 | 66 |