The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107320.1 | NP_001100790.1 | 7 - 8 | 938 | 97 |
XM_008771329.1 | XP_008769551.1 | 8 - 9 | 810 | 97 |