The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127542.1 | NP_001121014.1 | 3 - 4 | 428 | 93 |
GenBank mRNA | BC161922.1 | - | 3 - 4 | 414 | 93 |