The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175603.2 | NP_783193.2 | 11 - 11 | 1985 | 137 |
GenBank mRNA | BC093598.1 | - | 11 - 11 | 1985 | 137 |