Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam53b
Entrez Gene ID
309060
Gene Name
family with sequence similarity 53, member B
Gene Aliases
RGD1310799
Chromosome Location
Chr.1: 204714468 - 204805738 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759886.1 XP_008758108.1 1 - 2 2370 93

View Full Product Details