Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ethe1
Entrez Gene ID
292710
Gene Name
ethylmalonic encephalopathy 1
Gene Aliases
-
Chromosome Location
Chr.1: 81457008 - 81472061 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106234.1 NP_001099704.1 2 - 3 279 134
GenBank mRNA BC158848.1 - 2 - 3 239 134
FQ209618.1 - 2 - 3 250 134

View Full Product Details