Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Suds3
- Entrez Gene ID
-
360819
- Gene Name
- suppressor of defective silencing 3 homolog (S. cerevisiae)
- Gene Aliases
-
RGD1305986
- Chromosome Location
-
Chr.12: 45199436 - 45221504 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_341092.7 |
XP_341093.4 |
11 - 12 |
1038 |
100 |
View Full Product Details