Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx2
Entrez Gene ID
25130
Gene Name
syntaxin 2
Gene Aliases
Epim
Chromosome Location
Chr.12: 31324047 - 31360249 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012748.1 NP_036880.1 4 - 5 293 129
XM_006249249.2 XP_006249311.1 4 - 5 451 129
XM_006249250.2 XP_006249312.1 4 - 5 451 129
XM_006249253.2 XP_006249315.1 5 - 6 2197 129
XM_008769188.1 XP_008767410.1 5 - 6 2664 129
GenBank mRNA AY302700.1 - 4 - 5 293 129
L20823.1 - 4 - 5 293 129
L20888.1 - 4 - 5 293 129
L20889.1 - 4 - 5 293 129

View Full Product Details