Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps29
Entrez Gene ID
288666
Gene Name
vacuolar protein sorting 29 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.12: 39690069 - 39699127 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105932.1 NP_001099402.1 2 - 3 283 103
XM_006249362.2 XP_006249424.1 3 - 4 309 103
GenBank mRNA BC167055.1 - 3 - 4 270 103
FQ224287.1 - 3 - 4 291 103

View Full Product Details