Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tyw1
Entrez Gene ID
304423
Gene Name
tRNA-yW synthesizing protein 1 homolog (S. cerevisiae)
Gene Aliases
RGD1310420, Rsafd1
Chromosome Location
Chr.12: 29824990 - 29919240 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107137.1 NP_001100607.1 15 - 16 2078 110
GenBank mRNA BC168924.1 - 15 - 16 2060 110

View Full Product Details