Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Polr2j
Entrez Gene ID
288588
Gene Name
polymerase (RNA) II (DNA directed) polypeptide J
Gene Aliases
-
Chromosome Location
Chr.12: 23587222 - 23592774 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105921.1 NP_001099391.1 3 - 4 372 85

View Full Product Details