Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo21
Entrez Gene ID
360818
Gene Name
F-box protein 21
Gene Aliases
-
Chromosome Location
Chr.12: 44244457 - 44279044 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC158866.1 - 3 - 4 475 86

View Full Product Details