Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
288562
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr.12: 22177382 - 22194333 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105916.1 NP_001099386.1 16 - 17 1863 140
XM_006249116.2 XP_006249178.1 15 - 16 1815 140
XM_006249117.1 XP_006249179.1 16 - 17 1831 140
XM_006249118.1 XP_006249180.1 16 - 17 1834 140
XM_008769125.1 XP_008767347.1 15 - 16 1944 140
XM_008769126.1 XP_008767348.1 15 - 16 1774 140
XM_008769127.1 XP_008767349.1 15 - 16 1815 140
XM_008769128.1 XP_008767350.1 15 - 16 1815 140
GenBank mRNA BC166451.1 - 15 - 16 1928 140

View Full Product Details