Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem255b
Entrez Gene ID
290877
Gene Name
transmembrane protein 255B
Gene Aliases
Fam70b, RGD1566042
Chromosome Location
Chr.16: 81182478 - 81209429 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771387.1 XP_008769609.1 6 - 7 720 76
XM_008771388.1 XP_008769610.1 5 - 6 659 76
XM_008771389.1 XP_008769611.1 5 - 6 631 76

View Full Product Details