Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp2b4
Entrez Gene ID
29600
Gene Name
ATPase, Ca++ transporting, plasma membrane 4
Gene Aliases
PMCA4
Chromosome Location
Chr.13: 50091639 - 50153808 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005871.1 NP_001005871.1 9 - 10 1726 109
XM_008769446.1 XP_008767668.1 10 - 11 2109 109
XM_008769447.1 XP_008767669.1 10 - 11 2746 109
XM_008769448.1 XP_008767670.1 9 - 10 2073 109
XM_008769449.1 XP_008767671.1 10 - 11 2110 109
XM_008769450.1 XP_008767672.1 9 - 10 2074 109
GenBank mRNA U15408.1 - 9 - 10 1726 109

View Full Product Details