Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mael
Entrez Gene ID
364039
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
RGD1309333
Chromosome Location
Chr.13: 84412569 - 84452140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108857.1 NP_001102327.1 6 - 7 730 81

View Full Product Details