Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcsd1
Entrez Gene ID
360872
Gene Name
RCSD domain containing 1
Gene Aliases
RGD1304715
Chromosome Location
Chr.13: 83854650 - 83917037 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108349.1 NP_001101819.1 1 - 2 178 74
XM_006250181.2 XP_006250243.1 1 - 2 214 74

View Full Product Details