Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shcbp1l
Entrez Gene ID
289101
Gene Name
SHC SH2-domain binding protein 1-like
Gene Aliases
RGD1305609
Chromosome Location
Chr.13: 70852044 - 70882336 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105961.2 NP_001099431.1 5 - 6 1046 69
GenBank mRNA BC160910.1 - 5 - 6 1046 69

View Full Product Details