Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il20
Entrez Gene ID
681112
Gene Name
interleukin 20
Gene Aliases
-
Chromosome Location
Chr.13: 47644071 - 47651453 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143881.1 NP_001137353.1 2 - 3 443 85
XM_006249724.2 XP_006249786.1 3 - 4 458 85
GenBank mRNA DQ229286.1 - 2 - 3 230 85

View Full Product Details