Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo4
Entrez Gene ID
246247
Gene Name
flavin containing monooxygenase 4
Gene Aliases
-
Chromosome Location
Chr.13: 80685385 - 80703575 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144561.2 NP_653147.1 7 - 8 1137 165
NM_144562.1 NP_653148.1 5 - 6 742 165
XM_006250144.2 XP_006250206.1 7 - 8 1072 165
GenBank mRNA AF458416.1 - 7 - 8 1138 165
AF458417.1 - 5 - 6 742 165
BC081721.1 - 7 - 8 1127 165

View Full Product Details