Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc41a1
Entrez Gene ID
363985
Gene Name
solute carrier family 41 (magnesium transporter), member 1
Gene Aliases
-
Chromosome Location
Chr.13: 48607308 - 48627999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108855.1 NP_001102325.1 6 - 7 1870 97
XM_008769486.1 XP_008767708.1 5 - 6 993 97

View Full Product Details