Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spc25
Entrez Gene ID
295661
Gene Name
SPC25, NDC80 kinetochore complex component
Gene Aliases
RGD1307811, Spbc25
Chromosome Location
Chr.3: 55438534 - 55451934 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009654.1 NP_001009654.1 4 - 5 416 88
XM_006234288.2 XP_006234350.1 4 - 5 422 88
GenBank mRNA BC088213.1 - 4 - 5 416 88

View Full Product Details