Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mapk8ip1
Entrez Gene ID
116457
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
Gene Aliases
JIP1, Jip-1, Mapk8ip
Chromosome Location
Chr.3: 81295023 - 81304181 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053777.1 NP_446229.1 11 - 12 2239 123
GenBank mRNA AF108959.1 - 10 - 11 2177 123
AF109772.1 - 10 - 11 2086 123
AF109773.1 - 10 - 11 2149 123
AF109774.1 - 11 - 12 2239 123

View Full Product Details