Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oip5
Entrez Gene ID
499873
Gene Name
Opa interacting protein 5
Gene Aliases
RGD1564263
Chromosome Location
Chr.3: 111413662 - 111422319 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC167069.1 - 1 - 2 310 100

View Full Product Details