Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xirp2
Entrez Gene ID
311098
Gene Name
xin actin-binding repeat containing 2
Gene Aliases
Cmya3, LACS
Chromosome Location
Chr.3: 53563245 - 53658113 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201989.1 NP_973718.1 4 - 5 512 119
GenBank mRNA AF466694.1 - 4 - 5 512 119

View Full Product Details