Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd82
Entrez Gene ID
83628
Gene Name
Cd82 molecule
Gene Aliases
Kai1
Chromosome Location
Chr.3: 82324344 - 82368846 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031797.2 NP_113985.2 8 - 9 896 96
XM_006234597.2 XP_006234659.1 8 - 9 970 96
XM_006234598.1 XP_006234660.1 8 - 9 784 96
XM_008762039.1 XP_008760261.1 7 - 8 689 96
GenBank mRNA AF049882.1 - 8 - 9 810 96
BC072498.1 - 8 - 9 896 96

View Full Product Details