Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scn3a
Entrez Gene ID
497770
Gene Name
sodium channel, voltage-gated, type III, alpha subunit
Gene Aliases
Nav1.3, SCIII, Scn2a
Chromosome Location
Chr.3: 51530897 - 51643140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013119.1 NP_037251.1 11 - 12 2087 63
XM_008761914.1 XP_008760136.1 10 - 11 1890 63
XM_008761915.1 XP_008760137.1 11 - 12 1893 63
XM_008761916.1 XP_008760138.1 10 - 11 1891 63
XM_008761917.1 XP_008760139.1 10 - 11 1890 63
XM_008761918.1 XP_008760140.1 10 - 11 1890 63
XM_008761919.1 XP_008760141.1 10 - 11 1890 63
XM_008761920.1 XP_008760142.1 10 - 11 1890 63
GenBank mRNA Y00766.1 - 11 - 12 2087 63

View Full Product Details