Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
29231
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
Wdr39
Chromosome Location
Chr.3: 119671778 - 119677324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008766.1 NP_001008766.1 2 - 3 356 99
GenBank mRNA BC088474.1 - 2 - 3 356 99
FQ214368.1 - 2 - 3 379 99
FQ230934.1 - 2 - 3 370 99
FQ231672.1 - 2 - 3 349 99

View Full Product Details