Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epb42
Entrez Gene ID
362202
Gene Name
erythrocyte membrane protein band 4.2
Gene Aliases
Epb4.2
Chromosome Location
Chr.3: 112967095 - 112985318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108590.2 NP_001102060.1 1 - 2 131 110
GenBank mRNA BC168935.1 - 1 - 2 131 110

View Full Product Details