Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddrgk1
Entrez Gene ID
296162
Gene Name
DDRGK domain containing 1
Gene Aliases
RGD1309979
Chromosome Location
Chr.3: 123195580 - 123206828 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106512.1 NP_001099982.1 5 - 6 1066 123
GenBank mRNA FQ231424.1 - 5 - 6 690 123

View Full Product Details