Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rp9
Entrez Gene ID
363032
Gene Name
retinitis pigmentosa 9 (human)
Gene Aliases
RGD1559759, Rp9h
Chromosome Location
Chr.8: 23450111 - 23485759 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108756.1 NP_001102226.1 1 - 2 244 118

View Full Product Details