Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spc24
Entrez Gene ID
363028
Gene Name
SPC24, NDC80 kinetochore complex component
Gene Aliases
RGD1307631, Spbc24, Spc24-ps1
Chromosome Location
Chr.8: 22780639 - 22785720 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750448.3 XP_003750496.3 1 - 2 374 105

View Full Product Details