Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1e2
Entrez Gene ID
307091
Gene Name
aldo-keto reductase family 1, member E2
Gene Aliases
Akr1cl2, Akr1e1
Chromosome Location
Chr.17: 69365440 - 69379944 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008342.1 NP_001008343.1 2 - 3 279 73
GenBank mRNA BC086397.1 - 2 - 3 279 73
FQ212583.1 - 2 - 3 292 73

View Full Product Details