Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1e2
Entrez Gene ID
307091
Gene Name
aldo-keto reductase family 1, member E2
Gene Aliases
Akr1cl2, Akr1e1
Chromosome Location
Chr.17: 69365440 - 69379944 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008342.1 NP_001008343.1 5 - 6 593 131
XM_008771865.1 XP_008770087.1 4 - 5 496 131
GenBank mRNA BC086397.1 - 5 - 6 593 131
FQ212583.1 - 5 - 6 606 131

View Full Product Details