The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031641.1 | NP_001026811.1 | 8 - 9 | 946 | 70 |
XM_006253111.2 | XP_006253173.1 | 9 - 10 | 1071 | 70 | |
GenBank mRNA | BC098718.1 | - | 8 - 9 | 946 | 70 |