Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ska3
Entrez Gene ID
361047
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
RGD1307201
Chromosome Location
Chr.15: 38077903 - 38097100 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108379.1 NP_001101849.1 2 - 3 447 103
XM_006252074.1 XP_006252136.1 2 - 3 253 103
XM_006252075.1 XP_006252137.1 2 - 3 253 103
XM_008770758.1 XP_008768980.1 2 - 3 250 103
GenBank mRNA BC166461.1 - 2 - 3 224 103

View Full Product Details