The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108379.1 | NP_001101849.1 | 3 - 4 | 617 | 66 |
XM_006252074.1 | XP_006252136.1 | 3 - 4 | 423 | 66 | |
XM_006252075.1 | XP_006252137.1 | 3 - 4 | 423 | 66 | |
XM_008770758.1 | XP_008768980.1 | 3 - 4 | 420 | 66 | |
GenBank mRNA | BC166461.1 | - | 3 - 4 | 394 | 66 |