Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nop9
Entrez Gene ID
290235
Gene Name
NOP9 nucleolar protein
Gene Aliases
RGD1308396
Chromosome Location
Chr.15: 34431357 - 34439861 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106040.1 NP_001099510.1 7 - 8 1470 131

View Full Product Details