Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psme1
Entrez Gene ID
29630
Gene Name
proteasome (prosome, macropain) activator subunit 1
Gene Aliases
-
Chromosome Location
Chr.15: 34256071 - 34258947 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017264.3 NP_058960.2 1 - 2 148 66
GenBank mRNA BC060574.1 - 1 - 2 152 66
FQ216423.1 - 1 - 2 149 66
FQ219304.1 - 1 - 2 154 66
FQ220175.1 - 1 - 2 97 66
FQ224986.1 - 1 - 2 95 66
FQ226855.1 - 1 - 2 101 66
FQ228562.1 - - 103 66
FQ230622.1 - 1 - 2 95 66
FQ233635.1 - 1 - 2 101 66

View Full Product Details